C12H316CE3C1 Chemcia Scientific, LLC

8-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine

  • MDL number MFCD11846609
  • Cas Number : 1239648-74-8
  • Molecular Weight : 148.16861

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=559

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=559
Availability details for
Quantity
  • Availability
  • Add to Cart