C12H316CE3C3 Chemcia Scientific, LLC

5-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine

  • MDL number MFCD11846608
  • Cas Number : 1214901-53-7
  • Molecular Weight : 148.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl[1,2,4]triazolo[1,5-a]pyridin-2-ylamine
Inchi Code 1S/C7H8N4/c1-5-3-2-4-6-9-7(8)10-11(5)6/h2-4H,1H3,(H2,8,10)
Inchi Key VAXDBHYMMQDBLA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=561

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=561
Availability details for
Quantity
  • Availability
  • Add to Cart