C12H316CE3C4 Chemcia Scientific, LLC

Ethyl (6-Bromo-pyridin-2-ylamino)carbonothioylcarbamate

  • MDL number MFCD19441934
  • Cas Number : 1010120-59-8
  • Molecular Weight : 304.16695

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=562

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=562
Availability details for
Quantity
  • Availability
  • Add to Cart