C12H316CE3C7 Chemcia Scientific, LLC

6-Benzyl-5,6,7,8-tetrahydro-pyrido[4,3-d]pyrimidine

  • MDL number MFCD19441935
  • Cas Number : 1313712-15-0
  • Molecular Weight : 225.29575

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form Orange Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=565

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=565
Availability details for
Quantity
  • Availability
  • Add to Cart