C12H316CE3D9 Chemcia Scientific, LLC

2-N,N'-bis(tert-Butoxycarbonyl)amino-4,5,6,7-tetrahydro-benzo[b]thiophene-3-carboxylic acid methyl ester

  • MDL number MFCD19441939
  • Cas Number : 1313712-71-8
  • Molecular Weight : 411.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-[bis(tert-butoxycarbonyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Inchi Code 1S/C20H29NO6S/c1-19(2,3)26-17(23)21(18(24)27-20(4,5)6)15-14(16(22)25-7)12-10-8-9-11-13(12)28-15/h8-11H2,1-7H3
Inchi Key WVFDTFCCKCVWGQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=584

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=584
Availability details for
Quantity
  • Availability
  • Add to Cart