C12H316CE3DB Chemcia Scientific, LLC

2-tert-Butoxycarbonylamino-4,5,6,7-tetrahydro-benzo[b]thiophene-3-carboxylic acid methyl ester

  • MDL number MFCD19441941
  • Cas Number : 1313712-54-7
  • Molecular Weight : 311.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-[(tert-butoxycarbonyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Inchi Code 1S/C15H21NO4S/c1-15(2,3)20-14(18)16-12-11(13(17)19-4)9-7-5-6-8-10(9)21-12/h5-8H2,1-4H3,(H,16,18)
Inchi Key YOXOEVFUXYRJIK-UHFFFAOYSA-N

Description

Purity 90%
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=587

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=587
Availability details for
Quantity
  • Availability
  • Add to Cart