C12H316CE3E2 Chemcia Scientific, LLC

3-Amino-5-iodo-pyridin-4-ol hydrochloride

  • MDL number MFCD19441994
  • Cas Number : 1332581-59-5
  • Molecular Weight : 272.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-5-iodo-4-pyridinol hydrochloride
Inchi Code 1S/C5H5IN2O.ClH/c6-3-1-8-2-4(7)5(3)9;/h1-2H,7H2,(H,8,9);1H
Inchi Key ORCGMVUCDASTGU-UHFFFAOYSA-N

Description

Purity 97%
Salt Data HCl
Storage temp. 0~5
Physical Form Silver Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=472

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=472
Availability details for
Quantity
  • Availability
  • Add to Cart