C12H316CE3E4 Chemcia Scientific, LLC

4-Amino-3-hydroxy-benzonitrile hydrochloride

  • MDL number MFCD19442005
  • Cas Number : 1332605-83-0
  • Molecular Weight : 170.59964

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=468

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=468
Availability details for
Quantity
  • Availability
  • Add to Cart