C12H316CE3E5 Chemcia Scientific, LLC

8-Methyl-imidazo[1,2-a]pyridine hydrochloride

  • MDL number MFCD19442014
  • Cas Number : 10518-86-2
  • Molecular Weight : 168.62733

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Salt Data HCl
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=430

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=430
Availability details for
Quantity
  • Availability
  • Add to Cart