C12H316CE3E8 Chemcia Scientific, LLC

5-Methyl-imidazo[1,2-a]pyridine

  • MDL number MFCD06200991
  • Cas Number : 933-69-7
  • Molecular Weight : 132.16636

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Dark Brown Viscous Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=425

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=425
Availability details for
Quantity
  • Availability
  • Add to Cart