C12H316CE3EA Chemcia Scientific, LLC

3-Bromo-8-methyl-imidazo[1,2-a]pyridine

  • MDL number MFCD06496194
  • Cas Number : 866135-66-2
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-8-methylimidazo[1,2-a]pyridine
Inchi Code 1S/C8H7BrN2/c1-6-3-2-4-11-7(9)5-10-8(6)11/h2-5H,1H3
Inchi Key LPEITAUQQRNUAI-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=421

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=421
Availability details for
Quantity
  • Availability
  • Add to Cart