C12H316CE3EB Chemcia Scientific, LLC

3-Bromo-7-methyl-imidazo[1,2-a]pyridine

  • MDL number MFCD08056208
  • Cas Number : 56051-32-2
  • Molecular Weight : 211.06239

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=420

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=420
Availability details for
Quantity
  • Availability
  • Add to Cart