C12H316CE3F0 Chemcia Scientific, LLC

2-Dichloromethyl-6-methyl-imidazo[1,2-a]pyridine hydrochloride

  • MDL number MFCD19441993
  • Cas Number : 1332584-37-8
  • Molecular Weight : 251.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(dichloromethyl)-6-methylimidazo[1,2-a]pyridine hydrochloride
Inchi Code 1S/C9H8Cl2N2.ClH/c1-6-2-3-8-12-7(9(10)11)5-13(8)4-6;/h2-5,9H,1H3;1H
Inchi Key WXPCUIVLPILKJT-UHFFFAOYSA-N

Description

Purity 90%
Salt Data HCl
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=402

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=402
Availability details for
Quantity
  • Availability
  • Add to Cart