C12H316CE3F8 Chemcia Scientific, LLC

2-Ethyl-quinazolin-4-ol

  • MDL number MFCD16657449
  • Cas Number : 3137-64-2
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-ethyl-4-quinazolinol
Inchi Code 1S/C10H10N2O/c1-2-9-11-8-6-4-3-5-7(8)10(13)12-9/h3-6H,2H2,1H3,(H,11,12,13)
Inchi Key LUANCFNZZLKBOQ-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=684

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=684
Availability details for
Quantity
  • Availability
  • Add to Cart