C12H316CE3FC Chemcia Scientific, LLC

6-Nitro-8-bromo-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester hydrobromide

  • MDL number MFCD19442011
  • Cas Number : 1332605-92-1
  • Molecular Weight : 395.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 8-bromo-6-nitroimidazo[1,2-a]pyridine-2-carboxylate hydrobromide
Inchi Code 1S/C10H8BrN3O4.BrH/c1-2-18-10(15)8-5-13-4-6(14(16)17)3-7(11)9(13)12-8;/h3-5H,2H2,1H3;1H
Inchi Key RIXGMVGJZBEILK-UHFFFAOYSA-N

Description

Purity 96%
Salt Data HBr
Storage temp. 0~5
Physical Form Tan Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=351

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=351
Availability details for
Quantity
  • Availability
  • Add to Cart