C12H316CE3FE Chemcia Scientific, LLC

Chloro-acetic acid 2-(2-chloro-acetylamino)-benzyl ester

  • MDL number MFCD22565612
  • Cas Number : 1313712-59-2
  • Molecular Weight : 276.12122

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=685

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=685
Availability details for
Quantity
  • Availability
  • Add to Cart