C12H316CE3FF Chemcia Scientific, LLC

3-Bromo-7-chloro-6-iodo-quinolin-4-ol

  • MDL number MFCD15527297
  • Cas Number : 1021913-04-1
  • Molecular Weight : 384.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-7-chloro-6-iodo-4(1H)-quinolinone
Inchi Code 1S/C9H4BrClINO/c10-5-3-13-8-2-6(11)7(12)1-4(8)9(5)14/h1-3H,(H,13,14)
Inchi Key FLHMZLJOAWADLL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=702

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=702
Availability details for
Quantity
  • Availability
  • Add to Cart