C12H316CE41F Chemcia Scientific, LLC

2-tert-Butoxycarbonylamino-thiazole-4-carboxylic acid ethyl ester

  • MDL number MFCD09908086
  • Cas Number : 189512-01-4
  • Molecular Weight : 272.32517

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Beige Powder
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=773

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=773
Availability details for
Quantity
  • Availability
  • Add to Cart