C12H316CE423 Chemcia Scientific, LLC

3-Bromo-7-methyl-imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD20921526
  • Cas Number : 1352723-58-0
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-7-methylimidazo[1,2-a]pyridine-2-carboxylic acid
Inchi Code 1S/C9H7BrN2O2/c1-5-2-3-12-6(4-5)11-7(8(12)10)9(13)14/h2-4H,1H3,(H,13,14)
Inchi Key WJMIRFZVHMHRHU-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1195

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1195
Availability details for
Quantity
  • Availability
  • Add to Cart