C12H316CE424 Chemcia Scientific, LLC

2-Methyl-3H-quinazolin-4-one hydrochloride

  • MDL number MFCD22570184
  • Cas Number : 29378-39-0
  • Molecular Weight : 196.63788

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Salt Data HCl
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=778

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=778
Availability details for
Quantity
  • Availability
  • Add to Cart