C12H316CE427 Chemcia Scientific, LLC

4-Chloro-2-methoxy-5-nitro-benzoic acid

  • MDL number MFCD16873038
  • Cas Number : 68255-77-6
  • Molecular Weight : 231.59372

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=779

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=779
Availability details for
Quantity
  • Availability
  • Add to Cart