C12H316CE429 Chemcia Scientific, LLC

1-Methyl-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD00130001
  • Cas Number : 491-34-9
  • Molecular Weight : 147.22181

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Colourless Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=780

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=780
Availability details for
Quantity
  • Availability
  • Add to Cart