C12H316CE42E Chemcia Scientific, LLC

5,6,7,8-Tetrahydro-quinoline 1-oxide

  • MDL number MFCD01312585
  • Cas Number : 14631-48-2
  • Molecular Weight : 149.19412

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1367

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1367
Availability details for
Quantity
  • Availability
  • Add to Cart