C12H316CE42F Chemcia Scientific, LLC

4-Nitro-5,6,7,8-tetrahydroquinoline 1-oxide

  • MDL number MFCD20921528
  • Cas Number : 125162-98-3
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-nitro-5,6,7,8-tetrahydroquinoline 1-oxide
Inchi Code 1S/C9H10N2O3/c12-10-6-5-9(11(13)14)7-3-1-2-4-8(7)10/h5-6H,1-4H2
Inchi Key SWSDHCGZLPFBHU-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Powder
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=787

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=787
Availability details for
Quantity
  • Availability
  • Add to Cart