C12H316CE431 Chemcia Scientific, LLC

2,7-Dihydro-1H-azepine-3,6-dicarboxylic acid dimethyl ester

  • MDL number MFCD20921529
  • Cas Number : 1352723-54-6
  • Molecular Weight : 211.21941

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Beige Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=789

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=789
Availability details for
Quantity
  • Availability
  • Add to Cart