C12H316CE436 Chemcia Scientific, LLC

4-Fluoro-1,2-benzisoxazol-3-amine

  • MDL number MFCD09033911
  • Cas Number : 904815-05-0
  • Molecular Weight : 152.1291

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Beige Crystalline Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=795

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=795
Availability details for
Quantity
  • Availability
  • Add to Cart