C12H316CE43B Chemcia Scientific, LLC

N-Methyl-N-(5-fluoro-2-methylphenyl)amine

  • MDL number MFCD10000410
  • Cas Number : 881848-77-7
  • Molecular Weight : 139.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-N,2-dimethylaniline
Inchi Code 1S/C8H10FN/c1-6-3-4-7(9)5-8(6)10-2/h3-5,10H,1-2H3
Inchi Key NYLRQKXFBZTRMD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Dark brown liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=800

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=800
Availability details for
Quantity
  • Availability
  • Add to Cart