C12H316CE43C Chemcia Scientific, LLC

4-Chloro-2-methoxy-5-nitro-benzoic acid methyl ester

  • MDL number MFCD10574787
  • Cas Number : 109069-75-2
  • Molecular Weight : 245.62081

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Green Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=801

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=801
Availability details for
Quantity
  • Availability
  • Add to Cart