C12H316CE441 Chemcia Scientific, LLC

4-Bromo-N-methylphthalimide

  • MDL number MFCD00574083
  • Cas Number : 90224-73-0
  • Molecular Weight : 240.05767

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=940

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=940
Availability details for
Quantity
  • Availability
  • Add to Cart