C12H316CE442 Chemcia Scientific, LLC

3-(5-Chloro-thiophen-2-yl)-3-oxo-propionic acid ethyl ester

  • MDL number MFCD07783520
  • Cas Number : 188937-11-3
  • Molecular Weight : 232.68728

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=941

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=941
Availability details for
Quantity
  • Availability
  • Add to Cart