C12H316CE445 Chemcia Scientific, LLC

1-(3,5-Dimethylisoxazol-4-yl)ethanone

  • MDL number MFCD08701356
  • Cas Number : 35166-20-2
  • Molecular Weight : 139.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,5-dimethyl-4-isoxazolyl)ethanone
Inchi Code 1S/C7H9NO2/c1-4-7(5(2)9)6(3)10-8-4/h1-3H3
Inchi Key LNOJLNSJFRTBHL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1212

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1212
Availability details for
Quantity
  • Availability
  • Add to Cart