C12H316CE447 Chemcia Scientific, LLC

3,6,6-Trimethyl-1,5,6,7-tetrahydro-4H-indazol-4-one

  • MDL number MFCD00474387
  • Cas Number : 16315-16-5
  • Molecular Weight : 178.23588

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Pale Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=955

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=955
Availability details for
Quantity
  • Availability
  • Add to Cart