C12H316CE449 Chemcia Scientific, LLC

5-Bromo-4-nitro-1H-imidazole

  • MDL number MFCD13176330
  • Cas Number : 6963-65-1
  • Molecular Weight : 191.97229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2102

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2102
Availability details for
Quantity
  • Availability
  • Add to Cart