C12H316CE44B Chemcia Scientific, LLC

Indol-1-yl-acetic acid methyl ester

  • MDL number MFCD09834891
  • Cas Number : 39203-20-8
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 1H-indol-1-ylacetate
Inchi Code 1S/C11H11NO2/c1-14-11(13)8-12-7-6-9-4-2-3-5-10(9)12/h2-7H,8H2,1H3
Inchi Key FSWMODFYWAQRFP-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. 0~5
Physical Form Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=942

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=942
Availability details for
Quantity
  • Availability
  • Add to Cart