C12H316CE44E Chemcia Scientific, LLC

3-Methyl-1,5,6,7-tetrahydro-4H-indazol-4-one

  • MDL number MFCD00168632
  • Cas Number : 63446-38-8
  • Molecular Weight : 150.1817

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Powder
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=947

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=947
Availability details for
Quantity
  • Availability
  • Add to Cart