C12H316CE450 Chemcia Scientific, LLC

2H-Indazole-5-carboxylic acid methyl ester

  • MDL number MFCD22565613
  • Cas Number : 473416-12-5
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2H-indazole-5-carboxylate
Inchi Code 1S/C9H8N2O2/c1-13-9(12)6-2-3-8-7(4-6)5-10-11-8/h2-5H,1H3,(H,10,11)
Inchi Key LPLOEZPPYOSNEW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1213

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1213
Availability details for
Quantity
  • Availability
  • Add to Cart