C12H316CE452 Chemcia Scientific, LLC

5,6,7,8-Tetrahydro-1H-quinazoline-2,4-dione

  • MDL number MFCD13176330
  • Cas Number : 35042-48-9
  • Molecular Weight : 166.1811

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1791

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1791
Availability details for
Quantity
  • Availability
  • Add to Cart