C12H316CE454 Chemcia Scientific, LLC

5-Bromo-6-methyl-1,3-dihydropyrimidine-2,4-dione

  • MDL number MFCD00049045
  • Cas Number : 15018-56-1
  • Molecular Weight : 205.0118

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=959

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=959
Availability details for
Quantity
  • Availability
  • Add to Cart