C12H316CE459 Chemcia Scientific, LLC

2-Bromo-6-methyl-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD15529729
  • Cas Number : 1260678-93-0
  • Molecular Weight : 212.04997

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form White Needle-Like Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=964

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=964
Availability details for
Quantity
  • Availability
  • Add to Cart