C12H316CE45A Chemcia Scientific, LLC

(E)-2-(5-Bromo-2-(tosylimino)pyridin-1(2H)-yl)acetamide

  • MDL number MFCD20921552
  • Cas Number : 1005785-49-8
  • Molecular Weight : 384.25398

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=963

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=963
Availability details for
Quantity
  • Availability
  • Add to Cart