C12H316CE45D Chemcia Scientific, LLC

2-Bromo-7-methyl-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD19441943
  • Cas Number : 1352723-52-4
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-7-methyl-[1,2,4]triazolo[1,5-a]pyridine
Inchi Code 1S/C7H6BrN3/c1-5-2-3-11-6(4-5)9-7(8)10-11/h2-4H,1H3
Inchi Key WLGXBGDLTKSZAA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Powder
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=965

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=965
Availability details for
Quantity
  • Availability
  • Add to Cart