C12H316CE464 Chemcia Scientific, LLC

5,7-Dimethylpyrazolo[1,5-a]pyrimidine

  • MDL number MFCD01692984
  • Cas Number : 35149-38-3
  • Molecular Weight : 147.18103

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=931

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=931
Availability details for
Quantity
  • Availability
  • Add to Cart