C12H316CE468 Chemcia Scientific, LLC

6,7-Dihydro-1H-cyclopenta[d]pyrimidine-2,4(3H,5H)-dione

  • MDL number MFCD00466420
  • Cas Number : 5466-00-2
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6,7-dihydro-5H-cyclopenta[d]pyrimidine-2,4-diol
Inchi Code 1S/C7H8N2O2/c10-6-4-2-1-3-5(4)8-7(11)9-6/h1-3H2,(H2,8,9,10,11)
Inchi Key UPLURGSDLNKCSU-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=935

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=935
Availability details for
Quantity
  • Availability
  • Add to Cart