C12H316CE46B Chemcia Scientific, LLC

6-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-amine

  • MDL number MFCD11846607
  • Cas Number : 1239647-61-0
  • Molecular Weight : 148.16861

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form White Powder
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=928

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=928
Availability details for
Quantity
  • Availability
  • Add to Cart