C12H316CE474 Chemcia Scientific, LLC

5-Bromo-2,4-dichloro-6-methylpyrimidine

  • MDL number MFCD00504524
  • Cas Number : 56745-01-8
  • Molecular Weight : 241.90306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yelow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=978

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=978
Availability details for
Quantity
  • Availability
  • Add to Cart