C12H316CE47C Chemcia Scientific, LLC

2,3-Dihydrocinnolin-4(1H)-one hydrochloride

  • MDL number MFCD00156669
  • Cas Number : 137195-33-6
  • Molecular Weight : 184.62673

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HCl
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1002

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1002
Availability details for
Quantity
  • Availability
  • Add to Cart