C12H316CE485 Chemcia Scientific, LLC

6-Bromo-imidazo[1,2-a]pyrimidine-2-carboxylic acid ethyl ester

  • MDL number MFCD09838962
  • Cas Number : 944896-67-7
  • Molecular Weight : 270.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 6-bromoimidazo[1,2-a]pyrimidine-2-carboxylate
Inchi Code 1S/C9H8BrN3O2/c1-2-15-8(14)7-5-13-4-6(10)3-11-9(13)12-7/h3-5H,2H2,1H3
Inchi Key IMKBFCDPIFTPLH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1005

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1005
Availability details for
Quantity
  • Availability
  • Add to Cart