C12H316CE497 Chemcia Scientific, LLC

4-Iodo-3-methoxybenzenecarboxylic acid

  • MDL number MFCD09753732
  • Cas Number : 282087-44-9
  • Molecular Weight : 278.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-iodo-3-methoxybenzoic acid
Inchi Code 1S/C8H7IO3/c1-12-7-4-5(8(10)11)2-3-6(7)9/h2-4H,1H3,(H,10,11)
Inchi Key WUGQVHZHKSOODO-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Light Red Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1113

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1113
Availability details for
Quantity
  • Availability
  • Add to Cart