C12H316CE498 Chemcia Scientific, LLC

8-Bromoimidazo[1,2-a]pyridine-6-carboxylic acid hydrochloride

  • MDL number MFCD22570218
  • Cas Number : 1820666-77-0
  • Molecular Weight : 277.50622

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0~5
Physical Form Dark Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1111

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1111
Availability details for
Quantity
  • Availability
  • Add to Cart