C12H316CE49F Chemcia Scientific, LLC

Imidazo[1,2-a]pyrimidine hydrochloride

  • MDL number MFCD22416527
  • Cas Number : 6840-21-7
  • Molecular Weight : 155.58782

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Salt Data HCl
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1105

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1105
Availability details for
Quantity
  • Availability
  • Add to Cart